Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93229

Supelco

Ammonium hydrogen difluoride

for analysis EMPARTA®

Synonym(s):

Ammonium hydrogen difluoride, Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
MDL number:
UNSPSC Code:
12352302

mp

125 °C (lit.)

storage temp.

room temp

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93229.0521: 500 g, Minimum order quantity: 1
1.93229.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service