Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93696

Supelco

Sodium hexametaphosphate

EMPLURA®

Sign Into View Organizational & Contract Pricing

Synonym(s):
Sodium hexametaphosphate, Calgon, Phosphate glass, water soluble, Polyphosphate sodium salt, Sodium polyphosphate
CAS Number:
MDL number:
UNSPSC Code:
12352302

product line

EMPLURA®

storage temp.

room temp

InChI

1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2

InChI key

GYQBBRRVRKFJRG-UHFFFAOYSA-L

Biochem/physiol Actions

Intended for the cytologic demonstration of Naphthol AS-D Chloroacetate Esterase in blood, bone marrow films or tissue touch preparations.

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service