Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197220

Sigma-Aldrich

Bcl-2 Binding Peptide, Cell-Permeable

A cell-permeable peptide that binds to Bcl-2 with high affinity (IC₅₀ = 130 nM in a competition binding study) and induces apoptosis.

Synonym(s):

Bcl-2 Binding Peptide, Cell-Permeable, cpm-1285, Decanoyl-KN¹⁴⁰LWAAQRYGRELRRMSDEFEGSFKGL¹⁶⁵-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C153H240N44O42S
Molecular Weight:
3399.88
UNSPSC Code:
12352202

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable Bcl-2 binding peptide that induces apoptosis in Bcl-2 expressing cells with no apparent effect on normal peripheral lymphocytes. Also reported to diminish the growth of human myeloid leukemia in severe combined immunodeficient mice. The peptide is derived from the BH3 domain (residues 140-165, a death domain) of Bad and is modified to be cell-permeable by attaching a decanoyl moiety at the N-terminus.
A cell-permeable peptide that binds to Bcl-2 with high affinity (IC50 = 130 nM in a competition binding study) and induces apoptosis. Displays antitumor activity in cells overexpressing Bcl-2 with no apparent effect on normal peripheral lymphocytes. The peptide is derived from the BH3 domain (residues 140 - 165, a “death domain”) of Bad and is modified to be cell permeant by attaching a decanoyl moiety at the N-terminus.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Bcl-2
Product does not compete with ATP.
Reversible: no
Target IC50: 130 nM against Bcl-2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

CH₃(CH₂)₈CONH-Lys-Asn-Leu-Trp-Ala-Ala-Gln-Arg-Tyr-Gly-Arg-Glu-Leu-Arg-Arg-Met-Ser-Asp-Glu-Phe-Glu-Gly-Ser-Phe-Lys-Gly-Leu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Wang, J-L., et al. 2000. Cancer Res.60, 1498.
Kelekar, A., et al. 1997. Mol. Cell. Biol.17, 7040.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service