Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203630

Sigma-Aldrich

Bmi1-Ring1A E3 Ligase Inhibitor, PRT4165

Synonym(s):

PRT4165, 2-(3-Pyridinylmethylene)-1H-indene-1,3(2H)-dione; 2-(3-Pyridylmethylene)-1,3-indandione, NSC 600157

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9NO2
CAS Number:
Molecular Weight:
235.24
MDL number:
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

SMILES string

O=C1C(C(C2=C1C=CC=C2)=O)=CC3=CN=CC=C3

InChI

1S/C15H9NO2/c17-14-11-5-1-2-6-12(11)15(18)13(14)8-10-4-3-7-16-9-10/h1-9H

InChI key

OMHZFEWYVFWVLI-UHFFFAOYSA-N

Biochem/physiol Actions

PRT4165 is a potent inhibitor of PRC1 (polycomb-repressive complex 1)-mediated histone H2A ubiquitylation in vivo and in vitro. PRT4165 inhibits the in vitro histone H2A E3 ubiquitin ligase activity of PRC1 components RING1, RNF2, and a BMI1/RNF2 complex. PRT4165 also inhibits DNA double-strand breaks induced ubiquitylation of histone H2AX.
PRT4165 is a potent inhibitor of PRC1 (polycomb-repressive complex 1)-mediated histone H2A ubiquitylation in vivo and in vitro.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service