Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

204869

Sigma-Aldrich

Anti-Complement 3 Goat pAb

This Anti-Complement 3 Goat pAb is validated for use in Flow Cytometry

Synonym(s):

Anti-C3 Antibody, Complement 3 Detection Antibody, Goat Anti-Complement 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

biological source

goat

Quality Level

form

liquid

species reactivity

(baboon, not bovine, not canine, not chicken, not feline, guinea pig, hamster, horse, human, mouse, rabbit, rat, not sheep)

storage condition

OK to freeze

isotype

IgG

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
178467DR1001204903
Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

General description

Goat polyclonal antibody supplied as defibrinated, delipidized, and adsorbed (as needed) serum. Recognizes the C3 complement component.

Application

Flow Cytometry Immunoblotting Immunocytochemistry Immunoelectrophoresis (use undiluted)

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service