Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218

Sigma-Aldrich

Fuchsin Acid

HARLECO® For use in the Van Gieson connective tissue stain and Mallory′s aniline blue connective tissue stain

Synonym(s):

Fuchsin Acid, Acid Magenta, Acid Fuchsin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
585.54
Colour Index Number:
42685
MDL number:
UNSPSC Code:
41116130
EC Index Number:
221-816-5

Quality Level

form

solid

pH

3-4 (20 °C, 10 g/L in H2O)

mp

>130 °C (decomposition)

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c(ccc(c1)\C(=C3\C=C(C(=N)C(=C\3)C)[S](=O)(=O)[O-])\c2cc(c(cc2)N)[S](=O)(=O)O)N

InChI

1S/C20H19N3O9S3.2Na/c1-10-6-13(9-18(20(10)23)35(30,31)32)19(11-2-4-14(21)16(7-11)33(24,25)26)12-3-5-15(22)17(8-12)34(27,28)29;;/h2-9,23H,21-22H2,1H3,(H,24,25,26)(H,27,28,29)(H,30,31,32);;/q;2*+1/p-2

InChI key

WZRZTHMJPHPAMU-UHFFFAOYSA-L

General description

HARLECO® Fuchsin Acid
218-12 25g Glass Bottle

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
HARLECO® For use in the Van Gieson connective tissue stain and Mallory′s aniline blue connective tissue stain

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service