All Photos(1)

219537

Sigma-Aldrich

D-erythro-Sphingosine, Dihydro-, N-Acetyl- - CAS 13031-64-6 - Calbiochem

May be used as a negative control in studies of C₂ ceramide.

Synonym(s):
D-erythro-Sphingosine, Dihydro-, N-Acetyl- - CAS 13031-64-6 - Calbiochem, C₂ Dihydroceramide
Empirical Formula (Hill Notation):
C20H41NO3
CAS Number:
Molecular Weight:
343.54

Quality Level

assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

May be used as a negative control in studies of C2 ceramide. Its initial uptake appears to be slower than that of active C2 ceramide (Cat. No. 110145).
May be used as a negative control in studies of C2 ceramide. Its initial uptake appears to be slower than that of active C2 ceramide.

Packaging

5 mg in Plastic ampoule

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Primary Target
May be used as a negative control in studies of C2 ceramide
Cell permeable: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Powell, D.J., et al. 2003. Mol. cell Biol.23, 7794.
Hannun, Y.A. 1994. J. Biol. Chem.269, 3125.
Bielawska, A., et al. 1993. J. Biol. Chem.268, 26226.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service