Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

228

Sigma-Aldrich

Giemsa Stain Powder, Original Azure Blend

Harleco®, suitable for hematology, certified by the BSC

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

agency

certified by the BSC

form

solid

manufacturer/tradename

Harleco®

application(s)

diagnostic assay manufacturing
histology

hematology

shipped in

ambient

storage temp.

room temp

General description

HARLECO® Used for the staining of blood and bone marrow smears

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used in various staining protocols requiring Giemsa.
HARLECO® Used for the staining of blood and bone marrow smears

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service