MilliporeSigma
All Photos(1)

Documents

234149

Sigma-Aldrich

Collagen, Type I, Human Placenta

Sign Into View Organizational & Contract Pricing

Synonym(s):
Collagen, Type I, Human Placenta
CAS Number:

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
C8374C5533C7521
form

liquid

form

powder

form

powder

form

powder

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

-

storage condition

-

storage condition

-

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

Highly purified, native, type I collagen from human placenta. Useful as a gel or thin coating for the attachment of cells. Facilitates in vitro cultivation of cells and enhances cell-specific morphology and function. The recommended concentration is 0.2-2 µg/cm2 of growth surface.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM HCl.

Preparation Note

Further dilute with acetic acid (10 mM)

Other Notes

Bosco, M., et al. 1993. J. Cell Biol.120, 537.
Leavesley, D.I., et al. 1993. J. Cell Biol.121, 163.
Sidhu, J.S., et al. 1993. Arch. Biochem. Biophys.301, 103.
Streuli, C.H., et al. 1993. J. Cell Biol.120, 253.
Karsenty, G., et al. 1991. J. Biol. Chem.266, 24842.
Klasson, S.C., et al. 1986. Coll. Relat. Res.6, 397.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service