Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260684

Sigma-Aldrich

1-Deoxynojirimycin, Hydrochloride - CAS 73285-50-4 - Calbiochem

A specific glucosidase inhibitor including trimming glucosidases I and II, enzymes that sequentially remove the three glucose residues from precursor Glc3Man9-GlcNAc2 in N-linked glycan biosynthesis.

Synonym(s):

1-Deoxynojirimycin, Hydrochloride - CAS 73285-50-4 - Calbiochem, DNJ, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4 · xHCl
CAS Number:
Molecular Weight:
163.17 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 100 mM

shipped in

ambient

storage temp.

2-8°C

General description

A specific glucosidase inhibitor including trimming glucosidases I and II, enzymes that sequentially remove the three glucose residues from precursor Glc3Man9-GlcNAc2 in N-linked glycan biosynthesis.
Specific glucosidase inhibitor, including trimming glucosidases I and II, that sequentially removes the three glucose residues from precursor Glc3Man9GlcNAc2 in N-linked glycan biosynthesis.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Glucosidase 1, glucosidase 2
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Suh, K., et al. 1992. J. Biol. Chem. 267, 21671.
Fuhrmann, U., et al. 1985. Biochim. Biophys. Acta825, 95.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Suh et al.
The Journal of biological chemistry, 267(30), 21671-21677 (1992-10-25)
The role of glucosylated oligosaccharides in the biogenesis of the glycoprotein (G protein) of vesicular stomatitis virus was studied in PhaR2.7, a mouse lymphoma cell line deficient in glucosidase II activity. As expected, the great majority of cell-associated G protein
Inhibitors of oligosaccharide processing.
U Fuhrmann et al.
Biochimica et biophysica acta, 825(2), 95-110 (1985-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service