MilliporeSigma
All Photos(1)

Documents

264156-M

Millipore

Caspase-3 Inhibitor II

InSolution

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
InSolution Caspase-3 Inhibitor II, Z-D(OMe)E(Ome)VD(OMe)-FMK
Empirical Formula (Hill Notation):
C30H41FN4O12
Molecular Weight:
668.66

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

clear very faintly yellow

storage temp.

−20°C

General description

A potent, cell-permeable, and irreversible inhibitor of CPP-32/apopain, a member of the ICE/CED-3 family of cysteine proteases.

Biochem/physiol Actions

Primary Target
caspase-3, caspase-6, caspase-7, caspase-8, caspase-10

Warning

Toxicity: Irritant (B)

Sequence

Z-Asp(OCH₃)-Glu(OCH₃)-Val-Asp(OCH₃)-FMK

Physical form

A 5 mM (250 µg/75 µl) solution of Z-DEVD-FMK (Cat. No. 264155) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

single spot by TLC

Other Notes

Nicholson, D.W. 1996. Nat. Biotechnol.14, 297.
Schlegel, J., et al. 1996. J. Biol. Chem.271, 1841.0
Nicholson, D.W., et al. 1995. Nature376, 37.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service