328008
ERK Inhibitor II, Negative Control, FR180209
The ERK Inhibitor II, Negative Control, also referenced under CAS 1177970-73-8, controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
ERK Inhibitor II, Negative Control, FR180209, 5-(2-Phenyl-pyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-ol, Chk2 Inhibitor IV, Irfin1, Insulin Receptor Family Inhibitor, Aurora Kinase Inhibitor XII
About This Item
Quality Level
assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
green
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C18H12N6O/c25-18-12-10-13(19-20-17(12)21-22-18)15-14-8-4-5-9-24(14)23-16(15)11-6-2-1-3-7-11/h1-10H,(H2,20,21,22,25)
InChI key
QMBDONCHHMIWFJ-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Aurora B kinase
Packaging
Warning
Other Notes
Gao, Y., et al. 2013. Biochem. J.in press.
Ohori, M., et al. 2005. Biochem. Biophys. Res. Commun.336, 357.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service