Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341292

Sigma-Aldrich

PhosphoDetect Anti-FAK (pTyr³⁹⁷) Rabbit pAb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

biological source

rabbit

Quality Level

form

liquid

species reactivity

(Drosophila, chicken, frog, human, mouse, rat)

storage condition

OK to freeze

dilution

(Immunoblotting (1:1000)
Immunohistochemistry (heat pre-treatment required))

isotype

IgG

storage temp.

−70°C

General description

Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phosphorylation site, followed by immunoaffinity chromatography. Recognizes the ~125 kDa FAK protein phosphorylated on Tyr397.

Immunogen

a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr³⁹⁷ phosphorylation site of human FAK

Application

Immunoblotting (1:1000)Immunohistochemistry (heat pre-treatment required)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service