Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345795

Sigma-Aldrich

β1,3-Galactosidase, Xanthomonas manihotis, Recombinant, E. coli

Synonym(s):

β1,3-Galactosidase, Xanthomonas manihotis, Recombinant, E. coli, β-D-Galactoside Galactohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352202

recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥2000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

β-N-acetylglucosaminidase, α-fucosidase, α-galactosidase, β-glucosidase, α- and β-mannosidase, proteases, none detected

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, Xanthomonas manihotis β1,3-Galactosidase expressed in E. coli. Cleaves terminal β1,3-linked galactose residues on oligosaccharides. Has >100-fold kinetic preference for β1,3- over β1,6-linkages and 500-fold kinetic preference for β1,3- over β1,4-linkages.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave >95% of the β-D-galactose from 1 nmol Galβ1,3GlcNAcβ1,3Galβ1,4Glc-7-amino-4-methylcoumarin in 1 h at 37°C, pH 4.5 in a total volume of 10 µl.

Physical form

In 50 mM NaCl, 20 mM Tris-HCl, 100 µM EDTA, pH 7.5.

Other Notes

Taron, C., et al. 1995. Glycobiology 5, 603.
Wong-Madden, S.T., and Landry, D. 1995. Glycobiology 5, 19.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service