Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

354406

Sigma-Aldrich

Glutaredoxin-S2, E. coli

Glutaredoxin functions as a glutathione-dependent hydrogen donor for ribonucleotide reductase.

Synonym(s):

Grx1, grxA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

form

lyophilized

specific activity

≥175 μmole/min-mg (NADPH oxidized)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

protein purification: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Escherichia coli ... grxB(946926)

General description

Research area: CELL SIGNALING

Glutaredoxins (GRXs) are small thiol belonging to the Thioredoxin (TRX) superfamily. Different isoforms with varied functions have been identified in E.coli namely: glutaredoxin 1, glutaredoxin 2, and glutaredoxin 3. Native glutaredoxin-S2 isolated from E. coli. Glutaredoxin functions as a glutathione-dependent hydrogen donor for ribonucleotide reductase. It is useful as a general disulfide reductant for the in vitro study of protein folding mechanisms and has been demonstrated to work in conjunction with protein disulfide isomerase to enhance refolding of scrambled RNase A and RNase T1.

Application

Glutaredoxin-S2 has been used:
  • to catalyze the reduction of S-glutathionylated cysteine residues, to facilitate the detection of protein S-glutathionylation in FFPE HLTF-/-CDX sections.
  • for reduction of the protein sulfhydryl groups that were modified by glutathione for subsequent labeling and purification of glutathionylated mitochondrial proteins.

Biochem/physiol Actions

Glutaredoxins (Grxs) are a group of small redox proteins that play crucial roles in maintaining iron-sulfur metabolism and cellular redox homeostasis. These proteins function as thioltransferases , dehydroascorbate reductases, and transhydrogenases. They also participate in de-nitrosylation reactions and contribute to the conversion of cystine. Additionally, GRX2 plays a key role in neural and cardiac development. Low levels of GRX2 transcripts in humans may lead to conditions such as fibrosis, hypertrophy, and infarctions of the left ventricle.

Warning

Toxicity: Standard Handling (A)

Unit Definition

Units are defined using a standard HED assay.

Physical form

Lyophilized from 0.5% NH₄HCO₃.

Reconstitution

Reconstitute in 1 ml 50 mM Tris-HCl, 1 mM EDTA, pH 7.5 to yield a final concentration of 1 mg/ml.

Other Notes

Ruoppolo, M., et al. 1997. Biochemistry36, 12259.
Sun, C., et al. 1997. Protein Sci.6, 383.
Prinz, W.A., et al. 1997. J. Biol. Chem.272, 15661.
Holmgren, A. and Alund, F. 1995 Methods Enzymol.252, 283.
Hoog, J.O., et al. 1986. Gene43, 13.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein S-glutathionylation and the regulation of cellular functions
Mailloux RJ, et al.
Journal of Separation Science, 217-247 (2020)
J O Höög et al.
Gene, 43(1-2), 13-21 (1986-01-01)
Two DNA segments, together comprising 1147 bp and containing the glutaredoxin (GRX) gene, grx, from Escherichia coli K-12 were cloned and characterized in M13mp9. The gene was identified by hybridization with synthetic oligodeoxyribonucleotide probes corresponding to parts of the amino
Unique thiol metabolism in trypanosomatids: Redox homeostasis and drug resistance
Ali V, et al.
Advances in Parasitology, 117, 75-155 (2022)
M Ruoppolo et al.
Biochemistry, 36(40), 12259-12267 (1997-10-07)
Protein folding, associated with oxidation and isomerization of disulfide bonds, was studied using reduced and denatured RNase A (rd-RNase A) and mixed disulfide between glutathione and reduced RNase A derivative (GS-RNase A) as starting materials. Folding was initiated by addition
C Sun et al.
Protein science : a publication of the Protein Society, 6(2), 383-390 (1997-02-01)
Human glutaredoxin is a member of the glutaredoxin family, which is characterized by a glutathione binding site and a redox-active dithiol/disulfide in the active site. Unlike Escherichia coli glutaredoxin-1, this protein has additional cysteine residues that have been suggested to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service