Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

recombinant

expressed in E. coli

Quality Level

assay

≥90% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human GSK-3β fused at the N-terminus to a His•Tag sequence and expressed in E. coli. Glycogen synthase kinase-3 (GSK-3) is a serine/threonine kinase that was first characterized by its role in intermediary metabolism. GSK-3β is a dual-specificity kinase that is important role in insulin- and Wnt-mediated cellular signaling events. In Alzheimer′s disease GSK-3β is reported to affect microtubule stability via the phosphorylation of microtubule associated protein, Tau. Other known substrates include glycogen synthase, p90rsk, c-Jun, and CREB. MW: 53,000-56,000

Biochem/physiol Actions

1 µg GSK-3β maximally phosphorylates 100 ng Tau protein in 30 min in an in vitro kinase assay at 30°C, pH 7.5

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.1 M NH₄CO₃.

Reconstitution

Following reconstitution aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.
Reconstitute in sterile PBS to a final concentration of 1 mg/ml.

Other Notes

Harwood, A., and Braga, V.M. 2003. Nat. Cell Biol.5, 275.
Bhat, R.V., and Budd, S.L. 2002. Neurosignals11, 251.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service