Skip to Content
MilliporeSigma

370681

Sigma-Aldrich

OXE-R-G βγ Coupling Modulator, Gue1654 - Calbiochem

Synonym(s):

7-(Methylthio)-2-((2,2-diphenylacetyl)amino)benzo[1,2-d:4,3-dʹ]bisthiazole, N-(7-(Methylsulfanyl)[1,3]thiazolo[4,5-g][1,3]benzothiazol-2-yl)-2,2-diphenylacetamide

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

color

tan

solubility

DMSO: 25 mg/mL

General description

A membrane-permeant amidobenzobisthiazolo compound that modulates OXE-R-Gβγ coupling upon receptor stimulation via direct OXE-R interaction, but not preexisting OXE-R-Gαβ interaction. Shown to selectively inhibit OXE-R-Gβγ coupling-mediated, but not OXE-R-Gα-mediated, downstream signalings in HEK293 as well as primary human eosinophils and neutrophils (Effective Conc. 1 to 30 μM), displaying no inhibitory activity against cellular functions mediated by Gα or Gβγ when coupled to other GPCRs, including PGD2 receptor CRTH2 (DP2), IL8 receptors CXCR1/2, muscarinic receptor, and C5a receptor. Comparing to Gallein (Cat. No. 371708), Gue1654 does not exert its activity via direct Gβγ interaction.
A membrane-permeant amidobenzobisthiazolo compound that modulates OXE-R-Gβγ coupling upon receptor stimulation via direct OXE-R interaction, without affecting preexisting OXE-R-Gαβ interaction. Selectively inhibits OXE-R-Gβγ coupling-mediated, but not OXE-R-Gα-mediated, downstream cellular signalings (Effective Conc. 1 to 30 μM), displaying no inhibitory activity against cellular functions mediated by Gα or Gβγ when coupled to other GPCRs. Comparing to Gallein (Cat. No. 371708), Gue1654 does not exert its activity via direct Gβγ interaction.

Biochem/physiol Actions

OXE-R-Gβγ coupling

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service