MilliporeSigma
All Photos(1)

Documents

371780

Sigma-Aldrich

GP Antagonist-2A

The GP Antagonist-2A controls the biological activity of GP. This small molecule/inhibitor is primarily used for Neuroscience applications.

Sign Into View Organizational & Contract Pricing

Synonym(s):
GP Antagonist-2A, RPKPQQDWFDWDWM
Empirical Formula (Hill Notation):
C79H105N21O13S
Molecular Weight:
1588.88

Quality Level

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
553031346003344220
Sigma-Aldrich

371780

GP Antagonist-2A

Sigma-Aldrich

553031

RAGE Antagonist Peptide, RAP

Glucagon Receptor Antagonist II The Glucagon Receptor Antagonist II, also referenced under CAS 191034-25-0, controls the biological activity of Glucagon Receptor.

346003

Glucagon Receptor Antagonist II

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic)

General description

A peptide that selectively inhibits the activation of Gq by M1-muscarinic cholinergic receptors. Also inhibits activation by cholecystokinin receptor in pancreatic acini at 10 µM. Blocks angiotensin II-stimulated sis-inducing factor and STAT3 tyrosine phosphorylation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inhibits the activation of Gq by M1-muscarinic cholinergic receptors
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

H-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-D-Trp-Met-NH₂

Other Notes

Hunt, R.A., et al. 1999. Hypertension34, 603.
Tsunoda Y., et al. 1995. Biochem. Biophys. Res. Commun. 211, 648.
Mukai, H., et al. 1992. J. Biol. Chem.267, 16237.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service