Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

373278

Sigma-Aldrich

Hh Signaling Antagonist XII, MRT-83

The Hh Signaling Antagonist XII, MRT-83 controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Hh Signaling Antagonist XII, MRT-83, N-(2-Methyl-5-(3-(3,4,5-trimethoxybenzoyl)guanidino)phenyl)-(1,1ʹ-biphenyl)-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30N4O5
CAS Number:
Molecular Weight:
538.59
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O=C(NC(NC1=CC(NC(C2=CC=C(C3=CC=CC=C3)C=C2)=O)=C(C)C=C1)=N)C4=CC(OC)=C(C(OC)=C4)OC

General description

A cell-permeable acylguanidine compound that is shown to block Hh signaling and act as a potent, reversible and high-affinity Smo (Smoothened) antagonist. Displays 20-60 -fold greater potency than Cyclopamine (Cat. No. 239803) and competitively inhibits bodipy-cyclopamine binding to Smo (IC50 = 4.6 and 14 nM in HEK293-hSMO and HEK293-mSMO cells, respectively). Represses ShhN (N-myristoylated Shh) signaling (IC50 = 15 nM for Gli-dependent luciferase activity in Shh-light2 cells) and SAG-(Cat. Nos. 566660 & 566661) induced C3H10T1/2 cell differentiation (IC50 = 10 nM in an alkaline phosphatase activity assay). Inhibits ShhN (3 nM) and SAG (10 nM)-mediated proliferation of rat cerebellar granule cell precursors (IC50 ~ 3 and 6 nM, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Roudaut, H., et al. 2011. Mol. Pharmacol.79, 453.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service