Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

375110

Sigma-Aldrich

Hepatic Differentiation Inducer, SJA710-6

A cell-permeable imidazopyridinamine compound that selectively and potently induces differentiation of mesenchymal stem cells (MSCs) into hepatocyte-like cells (~47% at 5 µM).

Synonym(s):

Hepatic Differentiation Inducer, SJA710-6, 2-(4-Bromophenyl)-N-(4-fluorophenyl)-3-propyl-3H-imidazo[4,5-b]pyridin-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20BrFN4
CAS Number:
Molecular Weight:
439.32
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

BrC(C=C1)=CC=C1C2=NC3=C(N2CCC)N=C(NCC4=CC=C(F)C=C4)C=C3

General description

A cell-permeable imidazopyridinamine compound that selectively and potently induces differentiation of mesenchymal stem cells (MSCs) into hepatocyte-like cells (~47% at 5 µM). Within 14 days of treatment with SJA710-6, the differentiated cells exhibit typical morphological characteristics of hepatocytes and are able to store glycogen, secret albumin, produce urea, and uptake LDL. Also, shown to increase the mRNA levels of hepatocyte-specific genes, such as albumin, AFP, CK18, c-Met, CYP1A1, CYP2B1, HNF3β and FoxH1 .

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ouyang, J., et al. 2012. ChemMedChem7, In press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingfeng Ouyang et al.
ChemMedChem, 7(8), 1447-1452 (2012-06-21)
Mesenchymal stem cells (MSCs) are capable of self-renewal and multilineage differentiation. A periodic acid-Schiff (PAS) stain-based assay was developed to screen for small-molecule inducers of hepatic differentiation of bone marrow MSCs. 2-(4-Bromophenyl)-N-(4-fluorophenyl)-3-propyl-3H-imidazo[4,5-b]pyridin-5-amine (SJA710-6) was identified as a novel small molecule

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service