Skip to Content
MilliporeSigma

382184

Sigma-Aldrich

Histone Deacetylase 8, His•Tag®, Human Recombinant, S. frugiperda

Histone Deacetylase 8, His•Tag, Human Recombinant, S. frugiperda, is a full length HDAC8 fused to His-Tag at the C-terminus. Useful in the study of HDAC8 regulation and for inhibitor screening.

Synonym(s):

HDAC8, Human, Recombinant

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$748.00

$748.00


Check Cart for Availability

Select a Size

Change View
50 μG
$748.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$748.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

recombinant

expressed in insect cells (S. frugiperda)

Quality Level

assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥10 units/μg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
382180171539524733
assay

≥90% (SDS-PAGE)

assay

≥80% (SDS-PAGE)

assay

≥75% (SDS-PAGE)

assay

-

specific activity

≥10 units/μg protein

specific activity

≥50 units/μg protein

specific activity

≥270 nmol/min-mg protein

specific activity

≥0.25 U/μL

form

liquid

form

liquid

form

liquid

form

liquid

recombinant

expressed in insect cells (S. frugiperda)

recombinant

expressed in insect cells (S. frugiperda)

recombinant

-

recombinant

expressed in insect cells (S. frugiperda)

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

General description

Full length, recombinant, human histone deacetylase 8 (HDAC8) fused to at the C-terminus to a His•Tag sequence at and expressed in S. frugiperda insect cells using a baculovirus expression system. HDAC8 phosphorylated in vivo by PKA results in decreased activity, which leads to the hyperacetylation of histones H3 and H4. HDAC8 may play a role in one of the most frequent types of acute myeloid leukemia (AML). The active deacetylase is useful in the study of HDAC8 regulation and for inhibitor screening.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the the amount of enzyme required to deacetylate 1 picomol fluorescent peptide substrate [Arg-His-Lys(Ac)-Lys(Ac)] per min at 30°C.

Physical form

In 138 mM NaCl, 25 mM Tris-HCl, 10% glycerol, 0.05% TWEEN®-20 detergent, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Waltregny, D., et al. 2005. FASEB J.19, 966.
Buggy, J.J., et al. 2000. Biochem J. 350, 199

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service