Skip to Content
MilliporeSigma

400035

Sigma-Aldrich

Glucose Transporter Inhibitor II

The Glucose Transporter Inhibitor II controls the biological activity of Glucose Transporter.

Synonym(s):

Glucose Transporter Inhibitor II, ( Z)-5-(4-Hydroxy-3-trifluoromethyl-benzylidene)-3-(4,4,4-trifluoro-2-methyl-2-(2,2,2-trifluoroethyl)-butyl)-thiazolidine-2,4-dione

Slide 1 of 2
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C18H14F9NO3S
CAS Number:
Molecular Weight:
495.36
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥99% (HPLC)

form

powder

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1N(CC(CC(F)(F)F)(C)CC(F)(F)F)C(S/C1=C\C2=CC=C(O)C(C(F)(F)F)=C2)=O

General description

A cell-permeable thiazolidinedione compound that is shown to reversibly block glucose uptake in androgen-sensitive human prostate adenocarcinoma (LNCaP) cells transfected with GLUT1 (IC50 = 2 µM). Blocks GLUT3, GLUT4, and GLUT9-mediated transport at higher concentration (IC50 ≥ 5 µM). Binds to GLUT1 at a site that is distinct from the glucose binding site. Selectively reduces proliferation (IC50 = 2.0 µM) and induces apoptosis (IC50 = 2.5 µM) in LNCaP cells without affecting normal human prostate epithelial cells and mammary epithelial cells (~ 10 µM). Shown to elicit energy restriction-associated cellular responses in LNCaP cells, such as β-TrCP facilitated protein degradation, activation of AMP-activated kinase, and endoplasmic reticulum stress.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
glucose transporter
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Use only fresh DMSO.

Other Notes

Wang, D-S., et al. 2012, J. Med. Chem.55, 3827.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service