Skip to Content
MilliporeSigma

407697

Sigma-Aldrich

IR, β-Subunit, GST-Fusion, Human, Recombinant, S. frugiperda

Synonym(s):

Insulin Receptor β

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352202

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥80% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥50 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
325885325896171536
assay

≥80% (SDS-PAGE)

assay

-

assay

-

assay

≥95% (SDS-PAGE)

form

liquid

form

liquid

form

liquid

form

liquid

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

specific activity

≥50 units/mg protein

specific activity

≥30 units/mg protein

specific activity

≥1 units/mg protein

specific activity

-

General description

Recombinant, human IR β-subunit (amino acids 941-1343) fused to GST and expressed in S. frugiperda insect cells using a baculovirus expression system.

Physical form

In 50 mM Tris-HCl, 1 mM DTT, pH 7.5.

Other Notes

Al-Hasani, H., et al. 1994. FEBS Lett.349, 17.
One unit is defined as the amount of enzyme required to transfer 1 nmol phosphate to poly[Glu:Tyr] 4:1 substrate per min at 25°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service