Skip to Content
MilliporeSigma

419700

Sigma-Aldrich

Isopeptidase T, Rabbit

Synonym(s):

Ipase-T

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352202

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, rabbit isopeptidase T. A member of the higher molecular weight group of ubiquitin C-terminal hydrolases (UCHs) involved in the hydrolysis of isopeptide linkages of polyUb chains. Ipase-T activity is regulated by ubiquitin levels. Plays a major role in ubiquitin recycling and in the regulation of 26S proteasome activity. Recommended reaction buffer: 25 mM HEPES, 10 mM DTT, 25°C, pH 7.5. Pre-incubate the enzyme for 15 minutes with 10 mM DTT to achieve maximum activity.

Biochem/physiol Actions

Fully functional as measured using 5 nM Ipase-T and 1 µM Ub-AMC (Cat. No. 662075)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 50 mM HEPES, 1 mM DTT, pH 7.6.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Dang, L.C., et al. 1998. Biochemistry 37, 1868.
Melandri, F.D., et al. 1996. Biochemistry 35, 12893.
Stein, R.L., et al. 1995. Biochemistry 34, 12616.
Pickart, C.M., and Rose, I.A. 1986. J. Biol. Chem. 261, 10210.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service