419800
PARP Inhibitor IV, IQD
The PARP Inhibitor IV, IQD, also referenced under CAS 1188540, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Synonym(s):
PARP Inhibitor IV, IQD, 1,5-Dihydroxyisoquinoline, 5-Hydroxy-1(2H)-isoquinoline, 1,5-Isoquinolinediol
About This Item
Quality Level
assay
≥98% (NMR)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
DMSO: 30 mg/mL
0.1 M NaOH: 5 mg/mL
methanol: 8 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C9H7NO2/c11-8-3-1-2-7-6(8)4-5-10-9(7)12/h1-5,11H,(H,10,12)
Inchi Key
LFUJIPVWTMGYDG-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Poly(ADP-ribose) polymerase (PARS)
Packaging
Warning
Reconstitution
Other Notes
Dalamu, M., et al. 1996. Exp. Cell Res. 228, 14.
Zhang, J., et al. 1994. Science 263, 687.
Banasik, M., et al. 1992. J. Biol. Chem. 267, 1569.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service