Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420122

Sigma-Aldrich

JAK3 Inhibitor V

The JAK3 Inhibitor V, also referenced under CAS 932258, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK3 Inhibitor V, (2-Naphthylvinyl Ketone), ZM 449829

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

waxy solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C13H10O/c1-2-13(14)12-8-7-10-5-3-4-6-11(10)9-12/h2-9H,1H2

InChI key

FOTCGZPFSUWZBN-UHFFFAOYSA-N

General description

A breakdown product of JAK3 Inhibitor IV (Cat. No. 420121) with similar inhibitory potency (pIC50 = 6.8 for JAK3; 5.0 for EGF-R; 4.7 for JAK1). Also inhibits STAT-5 phosphorylation and T-cell proliferation.
A breakdown product of JAK3 Inhibitor-IV (Cat. No. 420121) with similar inhibitory potency (pIC50 = 6.8 for JAK3; 5.0 for EGF-R; 4.7 for JAK1). Also inhibits STAT-5 phosphorylation and T-cell proliferation.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no
pIC50 = 6.8 for JAK3; 5.0 for EGF-R; 4.7 for JAK1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Ethanol stock solutions are stable for up to 3 months at -20°C. Unstable in DMSO; reconstitute just prior to use.

Other Notes

Brown, G.R., et al. 2000. Bioorg. Med. Chem. Lett.10, 575.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service