Skip to Content
MilliporeSigma

444130

Mcl-1 Inhibitor, MIM1 - Calbiochem

Synonym(s):

4-((E)-((Z)-2-(cyclohexylimino)-4-methylthiazol-3(2H)-ylimino)methyl)benzene-1,2,3-triol, Mcl-1 Inhibitor Molecule 1, Mcl-1 Inhibitor, MIM1 - Calbiochem

Sign In to View Organizational & Contract Pricing

Select a Size

This product is sold exclusively by Millipore via offline channels. Contact Customer Service

About This Item

Linear Formula:
C17H21N3O3S
UNSPSC Code:
51111800
NACRES:
NA.28

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

(Yellow powder)

Quality Level

storage condition

OK to freeze

solubility

soluble (DMSO (50 mg/ml))

storage temp.

2-8°C

General description

A cell-permeable thiazolyliminopyrogallol that selectively neutralizes Mcl-1, but not Bcl-XL, antiapoptotic activity, by directly targeting Mcl-1 BH3-binding site, effectively competing against Bid BH3 sequence for Mcl-1 binding (IC50 = 4.8 µM) without affecting Bcl-XL interaction with Bad or Bid BH3 sequence (IC50 >50 µM). Shown to induce caspase 3/7 activation and cell death in Mcl-1-, but not Bcl-XL-, dependent B-ALL cultures (6% vs. 82% survival, respectively, [MIM1] = 10 µM). In comparison, ABT-737 (Cat. No. 197333) affects only Bcl-XL-, but not Mcl-1-, dependent B-ALL cultures (0% vs. 98% survival, respectively, [ABT-737] = 10 µM). Neither MIM1 nor ABT-737 significantly affects the survival of wild-type (81% and 79% survival in the presence of 10 µM respective inhibitor) or Bax-/-Bak-/- MEF (93% and 84% survival in the presence of 10 µM respective inhibitor).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service