Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444144

Sigma-Aldrich

MDM2 Antagonist II, NSC 66811

The MDM2 Antagonist II, NSC 66811, also referenced under CAS 6964-62-1, controls the biological activity of MDM2.

Synonym(s):

MDM2 Antagonist II, NSC 66811, 7-(Anilino(phenyl)-methyl)-2-methyl-8-quinolinol, MDM2 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C23H20N2O/c1-16-12-13-18-14-15-20(23(26)22(18)24-16)21(17-8-4-2-5-9-17)25-19-10-6-3-7-11-19/h2-15,21,25-26H,1H3

InChI key

WEENRMPCSWFMTE-UHFFFAOYSA-N

General description

A cell-permeable, non-peptidyl, quinolinol compound that binds MDM2 with high affinity (Ki = 120 nM) and disrupts MDM2-p53 interaction. Shown to dose-dependently induce cellular accumulation of p53, MDM2, and p21 in HCT-116 human colon cancer cell line with wild-type p53.
The MDM2 Antagonist II, NSC 66811, also referenced under CAS 6964-62-1, controls the biological activity of MDM2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MDM2
Product does not compete with ATP.
Target Ki: 120 nM against MDM2

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Lu, Y., et al. 2006. J. Med. Chem.49, 3759.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service