Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475250

Sigma-Aldrich

4-{[3ʹ,4ʹ-(Methylenedioxy)benzyl]amino}-6-methoxyquinazoline

Synonym(s):

4-{[3ʹ,4ʹ-(Methylenedioxy)benzyl]amino}-6-methoxyquinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O3
CAS Number:
Molecular Weight:
309.32
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

potency

230 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Potent and specific inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50 = 230 nM). Elevates the intracellular cGMP level without causing a change in the cAMP level in isolated porcine coronary arteries. Has no effect on other PDE isoenzymes.

Biochem/physiol Actions

Primary Target
CGMP-PDEV

Warning

Toxicity: Standard Handling (A)

Other Notes

Takase, Y., et al. 1994. J. Med. Chem.37, 2106.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service