Skip to Content
MilliporeSigma

475825

mitoNEET Inhibitor NL-1, Thiazolidinedione

The mitoNEET Inhibitor NL-1, Thiazolidinedione controls the biological activity of mitoNEET.

Synonym(s):

mitoNEET Inhibitor NL-1, Thiazolidinedione, 5-(3,5-di-tert-butyl-4-hydroxybenzyl)-4-hydroxythiazol-2(5H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C18H25NO3S
Molecular Weight:
335.46
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

mitoNEET Inhibitor NL-1, Thiazolidinedione, The mitoNEET Inhibitor NL-1, Thiazolidinedione controls the biological activity of mitoNEET.

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

DMSO: 100 mg/mL, clear, pale yellow

shipped in

ambient

storage temp.

−20°C

Quality Level

Disclaimer

Toxicity: Standard Handling (A)

General description

A cell-permeable, thiazolidinedione (TZD) that acts as an inhibitor of MitoNEET (IC50 = 0.9 µM) in competition with rosiglitazone and pioglitazone, in liver mitochondrial suspensions. In pharmacological studies, it was found to alter isolated mitochondrial complex I respiration with an IC50 of 2.4 µM and inhibited State III respiration up to 45%. Furthermore, this compound (6-25 µM) is shown to dose-dependently reduce lactate-dehydrogenase release in SH-SY5Y cultures, leading to a 40% decrease in rotenone-induced cell-death.

Other Notes

Geldenhuys, W.J., et al. 2009. Bioorg Med Chem Lett.3, 819.
Giles, R.G., et al. 2000. Tetrahedron56, 4531.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seunghee Lee et al.
Cell death & disease, 13(2), 127-127 (2022-02-10)
MitoNEET (mitochondrial protein containing Asn-Glu-Glu-Thr (NEET) sequence) is a 2Fe-2S cluster-containing integral membrane protein that resides in the mitochondrial outer membrane and participates in a redox-sensitive signaling and Fe-S cluster transfer. Thus, mitoNEET is a key regulator of mitochondrial oxidative
Aitor Martinez et al.
Molecular neurodegeneration, 19(1), 12-12 (2024-01-26)
Mitochondrial dysfunction and toxic protein aggregates have been shown to be key features in the pathogenesis of neurodegenerative diseases, such as Parkinson's disease (PD). Functional analysis of genes linked to PD have revealed that the E3 ligase Parkin and the
Kamalakannan Vijayan et al.
Cell chemical biology, 29(9), 1419-1433 (2022-06-24)
Prior to initiating symptomatic malaria, a single Plasmodium sporozoite infects a hepatocyte and develops into thousands of merozoites, in part by scavenging host resources, likely delivered by vesicles. Here, we demonstrate that host microtubules (MTs) dynamically reorganize around the developing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service