MilliporeSigma

5.00115

SAFC

Choline hydrogen tartrate

EMPROVE® ESSENTIAL, DAB 10, FCC

Sign Into View Organizational & Contract Pricing

Synonym(s):
Choline hydrogen tartrate, Choline bitartrate
Empirical Formula (Hill Notation):
C9H19NO7
CAS Number:
Molecular Weight:
253.25
EC Index Number:
201-763-4

Agency

DAB 10

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

pH

3.0-4.4 (20 °C, 50 g/L in H2O)

mp

147-152 °C

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/C5H14NO.C4H6O6/c1-6(2,3)4-5-7;5-1(3(7)8)2(6)4(9)10/h7H,4-5H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/q+1;/p-1/t;1-,2-/m.1/s1

InChI key

QWJSAWXRUVVRLH-LREBCSMRSA-M

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR14605.001171.00803
Choline hydrogen tartrate EMPROVE® ESSENTIAL, DAB 10, FCC

SAFC

5.00115

Choline hydrogen tartrate

Choline Bitartrate Pharmaceutical Secondary Standard; Certified Reference Material

Supelco

PHR1460

Choline Bitartrate

Choline chloride EMPROVE® ESSENTIAL, DAB 10

SAFC

5.00117

Choline chloride

L(+)-Tartaric acid powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF, E 334

SAFC

1.00803

L(+)-Tartaric acid

storage temp.

15-25°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

2-30°C

Quality Level

500

Quality Level

300

Quality Level

500

Quality Level

500

agency

DAB 10

agency

traceable to USP 1133536

agency

DAB 10

agency

BP, ChP, JP, NF, Ph. Eur.

reg. compliance

FCC

reg. compliance

-

reg. compliance

-

reg. compliance

-

product line

EMPROVE® ESSENTIAL

product line

-

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Choline hydrogen tartrate is an essential nutrient and typically grouped with B-complex vitamins.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service