Skip to Content
MilliporeSigma

5.04839

Sigma-Aldrich

(R)-CPP - CAS 126453-07-4 - Calbiochem

Synonym(s):

(R)-CPP - CAS 126453-07-4 - Calbiochem, (R)-3-(2-Carboxypiperazin-4-yl)propyl-1-phosphonic acid

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C8H17N2O5P
CAS Number:
Molecular Weight:
252.20
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 100 mM

storage temp.

2-8°C

General description

A highly potent antagonist slightly selective for NR2A over other subtype NMDA receptors (Ki = 0.041, 0.27, 0.63 and 1.99 µM for NMDA subtypes NR2A, NR2B, NR2C, and NR2D, respectively). Commonly used for various research in brain functions and disorders that neurotransmissions via NMDA receptors are involved.

Biochem/physiol Actions

Primary Target
NMDA receptors
Target Ki: 0.041, 0.27, 0.63 and 1.99 →M for NMDA subtypes NR2A, NR2B, NR2C, and NR2D, respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Murphy, R., et al. 2012. Psychopharmacol.219, 401.
Paoletti, P., et al. 2007. Curr. Opin. Pharmacol.7, 47.
Feng, B., et al. 2004. Br. J. Pharmacol.141, 508.
Lowe, A., et al. 1990. Neurosci. Lett.113, 315.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service