Skip to Content
MilliporeSigma

506173

Sigma-Aldrich

p38 MAP Kinase Inhibitor XI

Synonym(s):

p38 MAP Kinase Inhibitor XI

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C21H14F2N2O
Molecular Weight:
348.35
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light beige

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable naphthyridine N-oxide compound that acts as a potent, reversible p38α MAP kinase inhibitor (IC50 = 68 nM) and exhibits excellent selectivity among 54-kinases. It acts on the ATP-binding site plus an adjacent hydrophobic pocket. Exhibits desirable ADME (Absorption, Distribution, Metabolism, and Excretion) profile with good microsomal stability, low toxicity, and low reactivity towards cytochrome P450 isozymes. Shown to inhibit lipopolysaccharide-induced TNFα production in a dose-dependent manner (ED50 of 0.5 mg/kg).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Lumeras, W., et al. 2011. J. Med. Chem.54, 7899.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service