5.08018

Sigma-Aldrich

Herkinorin - CAS 862073-77-6 - Calbiochem

Synonym(s):
Herkinorin - CAS 862073-77-6 - Calbiochem, µ opioid receptor Agonist, Herkinorin
Empirical Formula (Hill Notation):
C28H30O8
CAS Number:
Molecular Weight:
494.53

storage conditions

-20C

assay

≥99% (HPLC)

Quality Level

form

solid

potency

45 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: soluble

General description

A potent agonist selective for µ opioid receptor (Ki = 45 nM) over κ opioid receptors (Ki = 184 nM). Widely used for pain research.

Biochem/physiol Actions

Primary Target
u opioid receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cui, X., et al. 2013. Eur.J. Pharmacol.702, 309.

Lamb, K., et al. 2012. Drug and Alcohol Dependence.121, 181.
Ji, F., et al. 2012. Brain Res.1490, 95.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.