513050
PDE10A Inhibitor, Cpd15
Synonym(s):
PDE10A Inhibitor, Cpd15, (6-chloro-3,8-dimethyl-1H-pyrazolo[3,4-b]quinolin-4-yl)(2-methylpyrimidin-5-yl)methanol, Phosphodiesterase 10A Inhibitor
About This Item
assay
≥98% (HPLC)
Quality Level
form
powder
potency
≤5 nM Ki
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 100 mg/mL
shipped in
wet ice
storage temp.
2-8°C
General description
Biochem/physiol Actions
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service