Skip to Content
MilliporeSigma

516569

PPARβ/δ Antagonist, PT-S58

The PPARβ/δ Antagonist, PT-S58 controls the biological activity of PPARβ/δ.

Synonym(s):

PPARβ/δ Antagonist, PT-S58, PPARβ Antagonist II, methyl 3-(N-(4-(tert-butylamino)-2-methoxyphenyl)sulfamoyl)thiophene-2-carboxylate, PPARδ Antagonist II

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C17H22N2O5S2
Molecular Weight:
398.50
MDL number:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark yellow

solubility

DMSO: 100 mg/mL, clear, amber-yellow

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, diaryl sulfonamide PPARβ/δ subtype-specific pure competitive antagonist with an IC50 of 98 nM. Antagonizes ligand binding without enabling coregulator interactions. Inhibits the PPARβ/δ specific agonist-induced transcriptional activity in cell cultures.

Packaging

Packaged under inert gas

Other Notes

Naruhn, S., et al. 2011, Mol. Pharm.80, 828.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service