Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

516571

Sigma-Aldrich

PPAR Agonist IX, GQ-16

The PPAR Agonist IX, GQ-16 controls the biological activity of PPAR. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

PPAR Agonist IX, GQ-16, ( Z)-5-(5-Bromo-2-methoxybenzylidene)-3-(4-methyl-benzyl)-thiazolidine-2,4-dione, (Z)-5-(5-Bromo-2-methoxybenzylidene)-3-(4-methyl-benzyl)-thiazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16BrNO3S
CAS Number:
Molecular Weight:
418.30
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

BrC1=CC(/C=C2SC(N(CC3=CC=C(C)C=C3)C\2=O)=O)=C(OC)C=C1

General description

A cell-permeable thiazolidinedione that acts as a PPARγ-selective agonist (Ki = 160 nM) and blocks Cdk-5-mediated Ser273 phosphorylation by stabilizing PPARγ β-sheet structure, exhibiting no detectable activity towards PPARα, PPARδ, or RXRα. Although a weaker PPAPγ agonist than Thiazolidinediones (TZDs) family rosiglitazone (Cat. No. 557366; Max. fold of transactivation induction = 8.57 and 24.8 with 10 µM respective compound in U-937 reporter assays), GQ-16 can be safely administered at a higher dosage (20 mg/kg/day; oral gavage) in mice to achieve similar in vivo efficacy as rosiglitazone (4 mg/kg/day; o.g.) in reversing high fat diet-induced insulin signaling defects without adverse side effects commonly seen with TZDs.such as edema and weight gain commonly seen with TZDs.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PPARγ
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Amato, A.A., et al. 2012. J. Biol. Chem.287, 28169.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service