Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525322

Sigma-Aldrich

PhosphoDetect Phosphotyrosine Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

Quality Level

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service