Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526511

Sigma-Aldrich

Phytohemagglutinin-M, Phaseolus vulgaris

Synonym(s):

Phytohemagglutinin-M, Phaseolus vulgaris

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.28
This product is sold exclusively by Millipore via offline channels. Contact Customer Service

description

Merck USA index - 14, 5429

Quality Level

form

lyophilized

storage condition

OK to freeze

solubility

soluble (Sterile redistilled H2O (2-10 mg/ml))

storage temp.

2-8°C

General description

T-cell mitogen that stimulates the proliferation of human lymphocytes and mouse splenic T-cells. PHA is a mixture of tetramers consisting of combinations of two subunits, E and L, with different biological activity. E-4 isolectin has low mitogenic activity and high erythroagglutinating activity. L-4 isolectin has high mitogenic and leukoagglutinating activities and low erythroagglutinating activity. Causes agglutination titers of 1:128 in tests that use a 2% solution of type A red blood cells. Inhibited by N-acetyl-D-galactosamine, peptides containing the sequence NANA-D-Gal-(1 → 3,4), GlcNAcα(1 → 2), or Man α(1 → 2)Man.

Packaging

Glass bottle

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service