Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526527

Sigma-Aldrich

PIM Kinase Inhibitor VIII, R8-T198wt

The PIM Kinase Inhibitor VIII, R8-T198wt controls the biological activity of PIM Kinase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

PIM Kinase Inhibitor VIII, R8-T198wt, NH₂-GGGRRRRRRRRGC~KKPGLRRRQT-CO₂H, cpm-P27 Kip1189-198, NH₂-GGGRRRRRRRRGC~KKPGLRRRQT-CO₂H, cpm-P27Kip1189-198

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C111H211N59O26S
Molecular Weight:
2820.31
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless (Use only fresh DMSO.)
water: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

General description

The human p27Kip1 C-terminus PIM-1 phosphorylation site- (Thr198) containing sequence is synthesized with an N-terminal Arg octamer/R8-containing sequence to facilitate its use as a cell-permeable, substrate-competitive PIM-1 inhibitor. Reported to retard human prostate carcinoma DU145-derived tumor growth in mice (100% vs 257% tumor growth on day 14 with or without 0.5 µmol/animal dosing on days 0 & 3; intratumoral injection) in vivo and completely prevents tumor expansion for up to 17 days (0.5 µmol/animal on days 0 & 5; intratumoral injection) when combined with a single Taxol (Cat. No. 580555) dosage (60 mg/kg; i.p.) on day zero. Studies using FITC-conjugated R8-T198wt confirms cellular delivery and PIM-1 interaction (KD = 323 nM), demonstrating also that R8-T198wt-induced cell cycle arrest and apoptosis in PIM-1-expressing DU145 cells are direct consequences of inhibition of PIM-1-mediated p27Kip1 Thr198 and Bad Ser112 phosphorylation, respectively.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Sequence

H-Gly-Gly-Gly-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Gly-Cys-Lys-Lys-Pro-Gly-Leu-Arg-Arg-Arg-Gln-Thr-OH

Physical form

Supplied as a trifluoroacetate salt

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Morishita, D., et al. 2011. J. Biol. Chem.286, 2681.
Morishita, D., et al. 2008. Cancer Res.68, 5076.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service