Skip to Content
MilliporeSigma

526560

PI 3-Kδ/γ Inhibitor XV, SW-14

The PI 3-Kδ/γ Inhibitor XV, SW-14 controls the biological activity of PI 3-Kδ/γ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-Kδ/γ Inhibitor XV, SW-14, 2-((4-amino-3-(3-fluoro-4-hydroxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)methyl)-5-methyl-3-o-tolylquinazolin-4(3H)-one, PI 3-K Inhibitor XV

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C28H22FN7O2
Molecular Weight:
507.52
UNSPSC Code:
12352200
Assay:
>95% (HPLC)
Form:
powder
Quality level:
Storage condition:
OK to freeze

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

PI 3-Kδ/γ Inhibitor XV, SW-14, The PI 3-Kδ/γ Inhibitor XV, SW-14 controls the biological activity of PI 3-Kδ/γ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

assay

>95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

Quality Level

Disclaimer

Toxicity: Standard Handling (A)

General description

A cell permeable, ATP competitive tolyl quinazolinone inhibitor of PI 3-Kδ/γ that displays 70-fold selectivity over PI 3-Kβ and 900-fold selectivity over PI 3-Kα in enzymatic assays with an IC50 = 9 nM for PI3Kδ and 21 nM for PI 3-Kγ and in a cell-based assay with an EC50 = 132 nM for PI 3-Kδ and 4.5 nM for PI 3-K γ. It elicits an inflammatory marker suppression effect in co-cultures of HUVEC and PBMCs stimulated with lipopolysacharide. Additionally, TNFα production decreases dose-dependently in the same cell lines treated with 370 nM to 10 µM of compound. SW14 exhibits a stronger effect compared to that of PI 3-Kδ inhibition (SW30) or pan-PI 3-K inhibition with different anti-inflammatory profile. In contrast to PI 3-Kα/β it also shows very little anti-proliferative effects even at the highest dose tested (10 µM) in HUVEC culture only.

Other Notes

Williams, O., et al. 2010. Chem Biol.17, 123.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service