Skip to Content
MilliporeSigma

528217

Sigma-Aldrich

Anti-PAI-1 Mouse mAb (MA-33H1F7)

This Anti-PAI-1 Mouse mAb (MA-33H1F7) is validated for use in Neutralization Studies for the detection of PAI-1.

Synonym(s):

Anti-PAI-1 Antibody, Mouse Anti-PAI-1, PAI-1 Detection Antibody

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

form

liquid

species reactivity

(human, mouse)

storage condition

OK to freeze

dilution

(Neutralization Studies)

isotype

IgG

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
528215IM29LST1684
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

General description

Purified monoclonal mouse antibody. Recognizes the ~50 kDa PAI-1 protein.

Immunogen

Human

Application

Neutralization Studies

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service