Skip to Content
MilliporeSigma

528284

Sigma-Aldrich

Polo-like Kinase Inhibitor III

The Polo-like Kinase Inhibitor III, also referenced under CAS 660868-91-7, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Polo-like Kinase Inhibitor III, 5-(5,6-Dimethoxy-1H-benzimidazol-1-yl)-3-((2-(trifluoromethyl)-benzyl)oxy)thiophene-2-carboxamide, Plk Inhibitor III

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C22H18F3N3O4S
CAS Number:
Molecular Weight:
477.46
MDL number:
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H18F3N3O4S/c1-30-16-7-14-15(8-17(16)31-2)28(11-27-14)19-9-18(20(33-19)21(26)29)32-10-12-5-3-4-6-13(12)22(23,24)25/h3-9,11H,10H2,1-2H3,(H2,26,29)

InChI key

JSKUWFIZUALZLX-UHFFFAOYSA-N

General description

A cell-permeable thiophene-benzimidazole compound that acts as a potent and reversible inhibitor against polo-like kinases (Ki against Plk1/2/3/4 = 4.8, 3.8, 8.0, and 163 nM , respectively), while exhibiting little or much reduced activity against 39 other commonly studied kinases. Preferentially inhibits the proliferation of a panel of 11 tumor cell lines (IC50 ≤0.7 µM) over normal human diploid fibroblasts (IC50 = 6.14 µM) in culture and is equally potent against the mdr-1-expressing MES-SA/DX5 and its parent non-drug resistant MES-SA uterine sarcoma cell line.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Johnson, E.F., et al. 2007. Biochemistry46, 9551.
Lansing, T.J., et al. 2007. Mol. Cancer Ther.6, 450.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service