Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539161

Sigma-Aldrich

Proteasome Inhibitor I

InSolution, ≥90%

Synonym(s):

InSolution Proteasome Inhibitor I, PSI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

storage temp.

−20°C

General description

Cell-permeable inhibitor of the chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells. Causes the accumulation of ubiquitinated proteins in neuronal cells. Prevents NF-κB activation in response to TNFα and okadaic acid through inhibition of IκBα degradation, thereby interfering with induction of iNOS in macrophages.

Biochem/physiol Actions

Primary Target
chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells
Product does not compete with ATP.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Z-Ile-Glu(OtBu)-Ala-Leu-CHO

Physical form

A 50 mM (5 mg/162 µl) solution of Proteasome Inhibitor I (Cat. No. 539160) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Griscavage, J.M., et al. 1996. Proc. Natl. Acad. Sci. USA93, 3308.
Mori, S., et al. 1995. J. Biol. Chem.270, 29447.
Figueiredo-Pereira, M.E., et al. 1994. J. Neurochem.63, 1578.
Traenckner, E.B., et al. 1994. EMBO J.13, 5433."

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

188.6 °F - (refers to pure substance)

flash_point_c

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service