Skip to Content
MilliporeSigma

539187

Sigma-Aldrich

Proteasome Inhibitor X, PI-083

The Proteasome Inhibitor X, PI-083, also referenced under CAS 6298-15-3, controls the biological activity of Proteasome. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Proteasome Inhibitor X, PI-083, NSC-45382

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C21H14ClN3O4S
CAS Number:
Molecular Weight:
439.87
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C21H14ClN3O4S/c22-18-19(21(27)16-6-2-1-5-15(16)20(18)26)24-13-8-10-14(11-9-13)30(28,29)25-17-7-3-4-12-23-17/h1-12,24H,(H,23,25)

InChI key

QRPKNPKHGNUFGK-UHFFFAOYSA-N

General description

A cell-permeable, reversible 20S proteasome inhibitor with IC50 values of 1 µM, 4.5 µM and 4.5 µM for CT-L, T-L and PGPH-L respectively. Exhibits CT-L (chymotrypsin-like) and cell proliferation inhibition selectively in ovarian, pancreatic, and breast cancer cells (T80-Hras, C7-Kras, and MCF-7, respectively) over their non-transformed counterparts (T80, C7, and MCF-10A), and induction of apoptosis with IC50 values of 3.7 µM in an MTT assay on bone marrow cells isolated from multiple myeloma patients in vitro. Shown to have anti-tumor activities in a nude mouse xenograft model implanted with MCF-7 and A549 cells in vivo when dosed at 1 mpk, leading to significant tumor growth inhibition of 84%. PI-083 results in 43, 12 and 34% inhibition of tumor CT-L, T-L and PGPH activities respectively, compared to those of tumors from vehicle-treated controls, while it does not inhibit liver proteasome activities.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lawrence, R., et al. 2010. Bioorg. Med. Chem.18, 5576.
Kazi, A., et al. 2009. Cell Cycle8, 1940.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service