Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539196

Sigma-Aldrich

Protease Inhibitor Cocktail Set X, EDTA-Free - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
solubility:
water: soluble
Pricing and availability is not currently available.

solubility

water: soluble

Quality Level

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service