MilliporeSigma
All Photos(1)

5500

Sigma-Aldrich

Light Diagnostics Rabies DFA II, FITC Conjugate, blend, clone 103.7 & 502.2

eCl@ss:
32161000

Quality Level

biological source

mouse

conjugate

FITC conjugate

clone

103.7, monoclonal
502.2, monoclonal

manufacturer/tradename

Chemicon®
Light Diagnostics

availability

available only in USA, Canada

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable

isotype

IgG2a

detection method

fluorometric

shipped in

wet ice

General description

Diagnostics Kit

Specificity

REACTIVITY: Clones react with lyssaviruses and can identify street and fixed rabies viruses as well as CVS-11 and HEP.

Application

INTENDED USE:

Analyte Specific Reagent. Not intended for diagnostic use.

DILUTION:

Optimal dilution is to be determined by the user.

Quality

PROCEDURE USED FOR QUALITY CONTROL TESTING OF CLONES:

1. Remove the acetone-fixed Positive and Negative Control impression slides from the freezer and allow to equilibrate to room temperature.

2. If uncoated slides are used, encircle each impressio

Storage and Stability

Sodium azide, present in this reagent can form potentially explosive metal azides with lead and copper pipes. Upon disposal, flush with large volumes of water to prevent build-up in plumbing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold as an Analyte Specific Reagent (ASR), for which the analytical and performance characteristics have not been established. According to current FDA regulations, those products labeled ASR can only be used by high-complexity labs, including those regulated by CLIA, public health labs, VA hospitals, and other diagnostic manufactures. ANY search or review for information on these products is considered by MILLIPORE to be a request for information by the searcher, and subsequently an acceptance on the part of the searcher of the limitations of the use of these products.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service