Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

555466

Sigma-Aldrich

Rho, GST-Fusion, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

capacity

≥400 mmol/mol binding capacity (GTPγS)

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human Rho fused to GST and expressed in E. coli. A small GTPase of the Ras superfamily. Regulates stress fiber formation in response to growth factor stimulation. Useful as a positive control in immunoblotting analysis and as an affinity precipitation reagent using immobilized glutathione resin.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 20 mM HEPES, 5 mM MgCl₂, 40% glycerol, 0.05% NaN₃, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Ridley, A. 1996. Curr. Biol. 6, 1256.
Ridley, A. and Hall, A. 1992. Cell 70, 389.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service