Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

555554

Sigma-Aldrich

Rho Kinase Inhibitor IV

The Rho Kinase Inhibitor IV controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Rho Kinase Inhibitor IV, (S)-(+)-2-Methyl-4-glycyl-1-(4-methylisoquinolinyl-5-sulfonyl)homopiperazine, 2HCl, H-1152, Glycyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N4O3S · 2HCl · xH2O
Molecular Weight:
449.40 (anhydrous basis)
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, reversible, and ATP-comptetive glycyl analog of Rho-Kinase Inhibitor (Cat. No. 555550) that inhibits ROCK with an improved selectivity (IC50 = 11.8 nM, >10 µM, >10 µM, 3.26 µM, 2.35 µM, and 2.57 µM for ROCKII, PKA, PKC, PKG, Aurora A, and CaMKII, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ROCK II
Product competes with ATP.
Reversible: yes
Target IC50: 11.8 nM against ROCK II

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tamura, M., et al. 2005. Biochim. Biophys. Acta, 1754, 245.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service