Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

557521

Sigma-Aldrich

Ro-31-8220

InSolution, ≥95%, protein kinase inhibitor

Synonym(s):

InSolution Ro-31-8220, 3-[1-[3-(Amidinothio)propyl-1H-indol-3-yl]-3-(1-methyl-1H-indol-3-yl)maleimide, Bisindolylmaleimide IX, Methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23N5O2S · xCH4O3S
Molecular Weight:
457.55 (free base basis)
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear orange-brown

shipped in

wet ice

storage temp.

−20°C

General description

A competitive, selective inhibitor for protein kinase C (IC50 = 10 nM) over protein kinase A (IC50 = 900 nM), Ca2+/calmodulin-dependent protein kinase (IC50 = 17 µM) and phosphorylase protein kinase. Inhibits TPA induced p47 phosphorylation in platelets (IC50 = 700 nM). Also inhibits CD3 downregulation in T cells (IC50 = 500 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target IC50: 10 nM against protein kinase C (PKC); 6.8 nM against GSK-3 in primary adipocytes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 5 mM (500 µg/181 µl) solution of Ro-31-8220 (Cat. No. 557520) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service